missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Brady™ GlobalMark™ Polyester Tapes

Durable outdoor-grade polyester with permanent adhesive.

$271.04 - $857.23

Specifications

Format Tape
Product Type Label Tape
For Use With (Application) Ideal for Stainless Steel and surfaces
Temperature Range (English) -40 to 230°F
Temperature Range (Metric) -40°C to +110°C
View More Specs

Products 34
Catalog Number Mfr. No. Color Length (English) Material Width (English) Price Quantity  
Catalog Number Mfr. No. Color Length (English) Material Width (English) Price Quantity  
19-201-152
View Documents
Brady™
76640
Green 100 ft. Polyester 2.25 in.
Each for $571.37
 
19-201-158
View Documents
Brady™
76621
Yellow 100 ft. Polyester 1.1 in.
Each for $401.96
 
19-201-157
View Documents
Brady™
76617
White 100 ft. Polyester 1.1 in.
Each for $401.96
 
19-201-159
View Documents
Brady™
76625
Red 100 ft. Polyester 1.1 in.
Each for $401.96
 
19-201-160
View Documents
Brady™
76629
Blue 100 ft. Polyester 1.1 in.
Each for $422.07
 
19-201-161
View Documents
Brady™
76633
Purple 100 ft. Polyester 1.1 in.
Each for $401.96
 
19-201-162
View Documents
Brady™
76637
Orange 100 ft. Polyester 1.1 in.
Each for $401.96
 
19-201-163
View Documents
Brady™
BRD76641
Green 100 ft. Polyester 1.1 in.
Each for $401.96
 
19-201-165
View Documents
Brady™
76649
Black 100 ft. Polyester 1.1 in.
Each for $433.72
 
19-201-164
View Documents
Brady™
76645
Brown 100 ft. Polyester 1.1 in.
Each for $401.96
 
19-201-167
View Documents
Brady™
76613
Clear 100 ft. Polyester 0.5 in.
Each for $271.04
 
19-201-168
View Documents
Brady™
76618
White 100 ft. Polyester 0.5 in.
Each for $271.04
 
19-201-170
View Documents
Brady™
76626
Red 100 ft. Polyester 0.5 in.
Each for $271.04
 
19-201-169
View Documents
Brady™
76622
Yellow 100 ft. Polyester 0.5 in.
Each for $271.04
 
19-201-175
View Documents
Brady™
76646
Brown 100 ft. Polyester 0.5 in.
Each for $271.04
 
19-201-174
View Documents
Brady™
76642
Green 100 ft. Polyester 0.5 in.
Each for $271.04
 
19-201-172
View Documents
Brady™
76634
Purple 100 ft. Polyester 0.5 in.
Each for $284.59
 
19-201-171
View Documents
Brady™
76630
Blue 100 ft. Polyester 0.5 in.
Each for $271.04
 
19-201-173
View Documents
Brady™
76638
Orange 100 ft. Polyester 0.5 in.
Each for $271.04
 
19-201-176
View Documents
Brady™
76650
Black 100 ft. Polyester 0.5 in.
Each for $271.04
 
19-201-177
View Documents
Brady™
76654
Gray 100 ft. Polyester 0.5 in.
Each for $271.04
 
19-201-182
View Documents
Brady™
76699
White 100 ft. Vinyl 1.125 in.
Each for $372.07
 
19-201-183
View Documents
Brady™
BRD76703
Yellow 100 ft. Vinyl 2.25 in.
Each for $375.55
 
19-201-193
View Documents
Brady™
76715
White 50 ft. Polyester 3.25 in.
Each for $554.43
 
19-201-192
View Documents
Brady™
76714
White 50 ft. Polyester 4 in.
Each for $642.20
 
19-201-194
View Documents
Brady™
76716
White 50 ft. Polyester 2.5 in.
Each for $429.68
 
19-201-197
View Documents
Brady™
76620
Yellow 100 ft. Polyester 2.25 in.
Each for $571.37
 
19-201-198
View Documents
Brady™
76712
Glow 50 ft. Polyester 2.5 in.
Each for $702.28
 
19-201-201
View Documents
Brady™
76713
Glow 50 ft. Polyester 1.1 in.
Each for $443.54
 
19-201-200
View Documents
Brady™
76721
Silver 100 ft. Metallized Polyester 1.1 in.
Each for $702.28
 
19-201-203
View Documents
Brady™
76722
Silver 100 ft. Metallized Polyester 0.5 in.
Each for $443.54
 
19-201-242
View Documents
Brady™
76612
Clear 100 ft. Polyester 1.1 in.
Each for $401.96
 
19-201-166
View Documents
Brady™
BRD76653
Gray 100 ft. Polyester 1.1 in.
Each for $401.96
 
19-108-014
View Documents
Brady™
76639
Green 100 ft. Polyester 4 in.
Each for $857.23
 
Description

Description

  • Low-halide content; ideal for marking stainless-steel pipes and surfaces
  • Ultrasmooth finish is excellent for mounting to rigid panels and very smooth surfaces
  • Two-year average outdoor durability in temperatures from 110°C to -40°C (230°F to -40°F); can be applied in temperatures as low as -18°C (0°F)

Order Info

Thickness: 3mil

Specifications

Specifications

Tape
Ideal for Stainless Steel and surfaces
-40°C to +110°C
Label Tape
-40 to 230°F
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'