Brady™ Label Maker Cartridge: Polyester, StainerBondz™
Label Maker Cartridge: StainerBondz™ Super Chemical Resistant Polyester (B-481) is designed for use in the slide staining process and other laboratory ID which requires superior chemical resistance.
$133.54 - $141.07
Specifications
Certifications/Compliance RoHS, ASTM D
Finish Matte White
Material Polyester
Sterility Non-sterile
For Use With (Application) Slide staining process and other laboratory identification which requires superior chemical resistance
View More Specs
Products 2
Catalog Number Mfr. No. Height (English) Width (English) Price Quantity  
Catalog Number Mfr. No. Height (English) Width (English) Price Quantity  
19-150-251
View Documents
Brady™
M194481
0.75 in. 0.9 in.
Each for $141.07
19-150-250
View Documents
Brady™
M195481
0.3 in. 0.9 in.
Each for $133.54
Description
Description
  • StainerBondz™ B-481 matte white polyester has a permanent acrylic adhesive and is thermal transfer printable with B30 series printers from Brady™
  • B-481 performs well in the commonly used H & E (hematoxilin and eosin) slide staining process
  • Excellent resistance to a wide variety of chemicals, oils and water
  • All-in-one cartridge includes label supply and R6400 Series ribbon
Specifications
Specifications
RoHS, ASTM D
Polyester
Slide staining process and other laboratory identification which requires superior chemical resistance
-112 to 266°F
40 lb./in.
0.9 in.
Matte White
Non-sterile
1 Ea.
-80 to -3°C
0.0033 in.
22.86 mm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'