missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Self-Laminating Tags
Self-Laminating Tags can be personalized by writing on special instructions, hazards or warnings.
Supplier: Brady™ 86456
Description
- B-851 Rigid Economy Polyester with brass grommets, exceeds OSHA's 50 lb pull force requirement for Tagout devices
- Textured write-on surface allows for authorized signatures, dates and other special instructions
- Seal and protect your written message with a clear adhesive overlay
- Withstands moisture, grease and dirt
Specifications
10/Pk. | |
Black, Yellow | |
Polyester | |
For marking on-the-spot warning of dangerous conditions | |
5.75 in. | |
0.01 in. | |
-40°F to 212°F | |
7.6 cm | |
0.93 cm |
OSHA | |
0.37 in. | |
CAUTION Do Not Remove This Tag | |
3 to 5 Years | |
14.6cm | |
-40°C to 100°C | |
3 in. | |
5.75 x 3 in. |