missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Self-Laminating Tags
Self-Laminating Tags can be personalized by writing on special instructions, hazards or warnings.
$42.28 - $48.35
Specifications
Quantity | 10/Pk. |
---|---|
Certifications/Compliance | OSHA |
Diameter (English) Grommet | 0.37 in. |
Material | Polyester |
For Use With (Application) | For marking on-the-spot warning of dangerous conditions |
Includes: 10 Nylon ties
Catalog Number | Mfr. No. | Color | Message | Dimensions (L x W) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Message | Dimensions (L x W) | Price | Quantity | |||
19-016467
|
Brady™
BRD65501 |
Black, Red, White | DANGER DO NOT OPERATE Do Not Remove This Tag It is Here To Protect My Life. Name/Date Do Not Remove This Tag/Remarks | 5.75 x 3 in. |
Pack of 10 Each for $42.28
|
|
|||
19-040-548
|
Brady™
81991 |
Black, Red, White | DANGER Do Not Remove This Tag/Remarks/See Other Side | 7 x 4 in. |
Pack of 10 Each for $48.35
|
|
|||
19040542
|
Brady™
86456 |
Black, Yellow | CAUTION Do Not Remove This Tag | 5.75 x 3 in. |
Pack of 10 Each for $43.85
|
|
|||
19-040-549
|
Brady™
86514 |
Yellow | CAUTION Do Not Remove This Tag/Remarks | 7 x 4 in. |
Pack of 10 Each for $48.35
|
|
|||
19-040-551
|
Brady™
86516 |
Black, Red, White | DANGER DO NOT OPERATE/ Signed By/Date/Phone Do Not Remove This Tag/Remarks | 7 x 4 in. |
Pack of 10 Each for $48.35
|
|
|||
19-040-552
|
Brady™
86517 |
Black, Red, White | DANGER Out Of Service/Signed By/Date/Phone Do Not Remove This Tag/Remarks | 7 x 4 in. |
Pack of 10 Each for $48.35
|
|
|||
19-040-804
|
Brady™
86461 |
Black, Red, White | DANGER DO NOT OPERATE Until this tag is removed by:/Signed By/Date/Phone Do Not Remove This Tag/Remarks | 5.75 x 3 in. |
Pack of 10 Each for $42.31
|
|
|||
Description
- B-851 Rigid Economy Polyester with brass grommets, exceeds OSHA's 50 lb pull force requirement for Tagout devices
- Textured write-on surface allows for authorized signatures, dates and other special instructions
- Seal and protect your written message with a clear adhesive overlay
- Withstands moisture, grease and dirt
Specifications
10/Pk. | |
0.37 in. | |
For marking on-the-spot warning of dangerous conditions | |
0.01 in. | |
-40°F to 212°F |
OSHA | |
Polyester | |
3 to 5 Years | |
-40°C to 100°C | |
0.93 cm |