Brady™ Self-Laminating Tags
Self-Laminating Tags can be personalized by writing on special instructions, hazards or warnings.
Supplier:  Brady™ 86461
 View more versions of this product
Catalog No. 19-040-804
Description
Description
  • B-851 Rigid Economy Polyester with brass grommets, exceeds OSHA's 50 lb pull force requirement for Tagout devices
  • Textured write-on surface allows for authorized signatures, dates and other special instructions
  • Seal and protect your written message with a clear adhesive overlay
  • Withstands moisture, grease and dirt
Specifications
Specifications
10/Pk.
Black, Red, White
Polyester
For marking on-the-spot warning of dangerous conditions
5.75 in.
0.01 in.
-40°F to 212°F
7.6 cm
0.93 cm
OSHA
0.37 in.
DANGER DO NOT OPERATE Until this tag is removed by:/Signed By/Date/Phone Do Not Remove This Tag/Remarks
3 to 5 Years
14.6cm
-40°C to 100°C
3 in.
5.75 x 3 in.