Brady™ U-Channel Sign Posts
Ideal for mounting and installing traffic signs
Supplier:  Brady™ 97204
 View more versions of this product
Catalog No. 19-054-905
Explore available promotions
Description
Description
  • Baked green enamel finish
  • Hot-rolled, high-tensile strength steel
  • 9.4mm (3/8 in.) pre-drilled holes, 2.54cm (1 in.) on center to accomodate most standard traffic signs
  • Weight: 2 lb. per linear foot
  • Sign mounting hardware and post driver sold separately
Specifications
Specifications
Steel
0.37 in.
8 ft.
U-channel Sign post
1 Ea.
Ideal for mounting and installation of traffic signs
2.4m
Green
Product Suggestions
Product Suggestions