missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Warning: Authorized Personnel Only Signs
Identify areas designated for certain personnel and direct facility traffic
Supplier: Brady™ 84191
Description
- Available in plastic, self sticking polyester and premium fiberglass materials
- Black on orange
- Language: english
- Available sizes (W x H): 10 x 7 in. or 14 x 10 in.
Specifications
ANSI Z535, OSHA 1910.145, PSTC-33 | |
1 Ea. | |
Used for safety signs, identification of fire alarms, extinguishers and emergency exits | |
17.7 cm | |
-40°F | |
10 in. | |
-40°C to 110°C | |
Warning Authorized Personnel Only | |
Black on Orange |
Polyester | |
Surface Mount | |
7 in. | |
230°F | |
0.006 to 0.01 in. | |
25.4 cm | |
-40°F to 230°F | |
Admittance Sign |