missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Warning: Electrical Hazard... Signs
Ensures safety at workplace
Supplier: Brady™ 69580
Description
- Black font on orange background
Specifications
ANSI Z535, OSHA 1910.145, PSTC-33, | |
1 Ea. | |
Premium Fiberglass | |
Industrial, utility, commercial and institutional environments for both indoor and outdoor use | |
193°F | |
0.1 in. | |
-40°C to 90°C | |
Warning Electrical Hazard Authorized Personnel Only | |
Black on Orange |
Fiberglass | |
9000 psi | |
Surface Mount | |
10 in. | |
-40°F | |
14 in. | |
-40°F to 193°F | |
Admittance Sign |