missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Brady™ Warning: Electrical Hazard... Signs
Ensures safety at workplace
$14.39 - $49.73
Specifications
Quantity | 1 Ea. |
---|---|
Mounting Type | Surface Mount |
Min. Temperature (English) | -40°F |
Displayed Text | Warning Electrical Hazard Authorized Personnel Only |
Color | Black on Orange |
Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Height (English) | Width (English) | Price | Quantity | |||
19035471
|
Brady™
22198 |
Plastic | 10 in. | 14 in. |
Each for $21.57
|
|
|||
19-035486
|
Brady™
69580 |
Fiberglass | 10 in. | 14 in. |
Each for $49.73
|
|
|||
19-035657
|
Brady™
84197 |
Polyester | 7 in. | 10 in. |
Each for $14.39
|
|
|||
19-035-919
|
Brady™
84198 |
Polyester | 10 in. | 14 in. |
Each for $36.88
|
|
|||
Description
- Black font on orange background
Specifications
1 Ea. | |
-40°F | |
Black on Orange |
Surface Mount | |
Warning Electrical Hazard Authorized Personnel Only |