missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BrandTech™ BRAND™ seripettor™ pro Bottletop Dispensers
Designed to handle a wider range of liquids than the regular seripettor
Supplier: BrandTech™ BRAND™ 4720450
Includes: Each seripettor pro bottletop dispenser is supplied with discharge tube, filling tube, spare dispensing catridge, mounting tool, two polypropylene adapters (GL45/33 and GL45/38), operating manual, and one-year warranty.
Description
Easily-replaceable wiping seal dispensing cartridge is especially suitable for liquids that tend to form crystals
- Quick-change cartridge makes service a breeze and keeps life-time service costs low
- Pump assembly with corrosion resistant spring has opaque upper sleeve that protects liquid from UV-light during dispensing operations
- Telescoping filling tube adjusts to fit many bottles without cutting
- Threaded safety cap attaches and removes with a quick twist
- Spring-loaded piston permits one-handed, semi-automatic operation
Liquids unsuitable for the seripettor pro include, but are not limited to:
- Hydrofluoric acid, or other chemicals that attack high-density polyethylene,PTFE, ETFE, FEP, PFA, alumina-ceramic, platinum-iridium, and borosilicate glass
- Carbon disulfide
- Density exceeding 2.2g/cm
- Viscosity exceeding 300mm2/sec (2mL instrument), 150mm2/s (10mL instrument), 75mm2/s (25mL instrument)
- Vapor pressure up to 500mbar
Agar culture media to 140°F (60°C).
Order Info
Replaceable piston and cylinder are permanently connected as a cartridge module for safety. If piston, cylinder, or seal are damaged, simply replace cartridge in seconds without tools.
Specifications
≤±1.2% | |
Up to 2.2 g/cm3 | |
Discharge tube, telescoping filling tube, spare dispensing cartridge, mounting tool, GL45/33mm and GL45/38mm polypropylene adapters, operating manual | |
≤0.2% | |
15°C to 40°C | |
75 mm2/sec. |
Seripettor Pro Bottletop Dispenser | |
2.5 to 25 mL | |
0.5 mL | |
59°F to 104°F | |
Up to 500mbar | |
1 Year |