missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BrandTech™ BRAND™ seripettor™ pro Bottletop Dispensers

Designed to handle a wider range of liquids than the regular seripettor

$903.70 - $1282.85

Specifications

Product Type Seripettor Pro Bottletop Dispenser
Accuracy ≤±1.2%
Density Up to 2.2 g/cm3
Includes Discharge tube, telescoping filling tube, spare dispensing cartridge, mounting tool, GL45/33mm and GL45/38mm polypropylene adapters, operating manual
Precision ≤0.2%
View More Specs

Products 3
Catalog Number Mfr. No. Volume (Metric) Increments Viscosity Rating Price Quantity  
Catalog Number Mfr. No. Volume (Metric) Increments Viscosity Rating Price Quantity  
13-688-400
View Documents
BrandTech™ BRAND™
4720450
2.5 to 25 mL 0.5 mL 75 mm2/sec.
Each for $1,282.85
 
13-688-399
View Documents
BrandTech™ BRAND™
4720440
1 to 10 mL 0.2 mL 150mm2/sec.
Each for $903.70
 
03-840-011
View Documents
BrandTech™ BRAND™
4720420
0.2 to 2 mL 0.04 mL 1000mm2/sec.
Each for $903.70
 
Description

Description

Easily-replaceable wiping seal dispensing cartridge is especially suitable for liquids that tend to form crystals

  • Quick-change cartridge makes service a breeze and keeps life-time service costs low
  • Pump assembly with corrosion resistant spring has opaque upper sleeve that protects liquid from UV-light during dispensing operations
  • Telescoping filling tube adjusts to fit many bottles without cutting
  • Threaded safety cap attaches and removes with a quick twist
  • Spring-loaded piston permits one-handed, semi-automatic operation

Liquids unsuitable for the seripettor pro include, but are not limited to:

  • Hydrofluoric acid, or other chemicals that attack high-density polyethylene,PTFE, ETFE, FEP, PFA, alumina-ceramic, platinum-iridium, and borosilicate glass
  • Carbon disulfide
  • Density exceeding 2.2g/cm
  • Viscosity exceeding 300mm2/sec (2mL instrument), 150mm2/s (10mL instrument), 75mm2/s (25mL instrument)
  • Vapor pressure up to 500mbar
Temperature of instrument and reagents must be between 59° and 104°F (15° and 40°C).
Agar culture media to 140°F (60°C).

Order Info

Replaceable piston and cylinder are permanently connected as a cartridge module for safety. If piston, cylinder, or seal are damaged, simply replace cartridge in seconds without tools.

Specifications

Specifications

Seripettor Pro Bottletop Dispenser
Up to 2.2 g/cm3
≤0.2%
15°C to 40°C
1 Year
≤±1.2%
Discharge tube, telescoping filling tube, spare dispensing cartridge, mounting tool, GL45/33mm and GL45/38mm polypropylene adapters, operating manual
59°F to 104°F
Up to 500mbar
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'