missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BrandTech™ BRAND™ Centrifuge Tube Mini-Cooler
Designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench.
Supplier: BrandTech™ BRAND™ 114930
Description
- Manufactured from durable polycarbonate
- Filled with non-toxic gel
- Hold twelve 0.5 to 2.0mL tubes
- Provided individually
Specifications
Centrifuge tube | |
Polycarbonate | |
Red | |
0.5 to 2 mL Tubes |
Maintains 0°C temperature for 60 min. | |
Mini Cooler | |
0.5 to 2 mL |