missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
BrandTech™ BRAND™ Centrifuge Tube Mini-Cooler
Designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench.
$682.24 - $682.24
Specifications
For Use With (Equipment) | Centrifuge tube |
---|---|
Material | Polycarbonate |
Type | Mini Cooler |
Capacity (Metric) | 0.5 to 2 mL |
Holds | 0.5 to 2 mL Tubes |
Catalog Number | Mfr. No. | Color | Description | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Description | Price | Quantity | |||||
14-380-801
|
BrandTech™ BRAND™
114935 |
Yellow | Maintains −20°C temperature for 60 min. |
Each for $682.24
|
|
|||||
14-380-800
|
BrandTech™ BRAND™
114930 |
Red | Maintains 0°C temperature for 60 min. |
Each for $682.24
|
|
|||||
Description
- Manufactured from durable polycarbonate
- Filled with non-toxic gel
- Hold twelve 0.5 to 2.0mL tubes
- Provided individually
Specifications
Centrifuge tube | |
Mini Cooler | |
0.5 to 2 mL Tubes |
Polycarbonate | |
0.5 to 2 mL |