missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

BrandTech™ BRAND™ Centrifuge Tube Mini-Cooler

Designed to protect a wide range of solutions (enzymes, DNA, RNA, cell suspensions) by helping to maintain freezer temperatures on the lab bench.

Supplier:  BrandTech™ BRAND™ 114935

 View more versions of this product

Catalog No. 14-380-801


Explore available promotions

Description

Description

  • Manufactured from durable polycarbonate
  • Filled with non-toxic gel
  • Hold twelve 0.5 to 2.0mL tubes
  • Provided individually

Specifications

Specifications

Centrifuge tube
Polycarbonate
Yellow
0.5 to 2 mL Tubes
Maintains −20°C temperature for 60 min.
Mini Cooler
0.5 to 2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions