Branson Ultrasonics™ Bransonic™ Ultrasonic Cleaner Model B200
Compact and stylish, with convenience of plug-in-anywhere operation. Branson Ultrasonics Bransonic™ Ultrasonic Cleaner Model B200 designed to clean jewelry and optical pieces quickly and effectively with ultrasonic cleaning ability.
Supplier: Branson Ultrasonics™ 100951010
Description
- Five-minute timer shuts off automatically
- Stainless-steel tank
- Sealed within an impact-resistant plastic housing
Specifications
5 min. | |
60 Hz | |
15 oz. | |
8.8 cm | |
To clean instrument and clock parts, small geological samples, small electrical/electronic components, metal and plastic machined parts, recorder and drafting pens | |
8.75 in. | |
22.2 cm | |
B200 | |
1.36 kg | |
11.4 cm |
Plastic Housing, Stainless Steel Tank | |
120 V | |
400 mL | |
120V 60Hz | |
Cover and parts basket | |
6.5 in. | |
16.5 cm | |
3 lb. | |
3.5 in. |