missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ BrightComp eBeads™ Compensation Bead Kits DFS Item

BrightComp eBeads compensation beads provide a consistent, accurate, and simple-to-use technique for setting flow cytometry compensation when using fluorescent protein-expressing samples.

Supplier:  Invitrogen™ A10514

 View more versions of this product

Catalog No. A10514


Explore available promotions

Description

Description

BrightComp eBeads compensation beads provide a consistent, accurate, and simple-to-use technique for setting flow cytometry compensation when using fluorescent protein-expressing samples.

  • Three levels of intensity, perfect for any samples no matter the level of expression
  • Near-identical, spectrally-matched dye to fluorescent protein
  • Consistent, reliable performance
Accurate compensation is crucial for effective multicolor analysis in flow cytometry. Each kit provides polystyrene microspheres with near-identical, spectrally-matched fluorescent protein dye as well as negative control beads in a single dropper bottle.
Specifications

Specifications

Store at 2°C to 8°C. DO NOT FREEZE.
Compensation Beads
Green
Flow Cytometry
488/525
GFP BrightComp eBeads™ Compensation Bead Kit
Room Temperature
25 tests
eBeads™
Flow Cytometer
Videos
SDS
Documents

Documents

Promotions

Promotions