Invitrogen™ BrightComp eBeads™ Compensation Bead Kits DFS Item
BrightComp eBeads compensation beads provide a consistent, accurate, and simple-to-use technique for setting flow cytometry compensation when using fluorescent protein-expressing samples.
$473.00 - $473.00
Specifications
For Use With (Application) Flow Cytometry
Product Type Compensation Beads
Quantity 25 tests
View More Specs
Products 1
Catalog Number Mfr. No. Description Price Quantity  
Catalog Number Mfr. No. Description Price Quantity  
A10514 DFS Item
View Documents
Invitrogen™
A10514
GFP BrightComp eBeads™ Compensation Bead Kit
Pack for $473.00
Description
Description
BrightComp eBeads compensation beads provide a consistent, accurate, and simple-to-use technique for setting flow cytometry compensation when using fluorescent protein-expressing samples.
• Three levels of intensity, perfect for any samples no matter the level of fluorescent protein expression
• Near-identical, spectrally-matched dye to native protein
• Consistent, reliable performance
Accurate compensation is crucial for effective multicolor analysis in flow cytometry. Each kit provides polystyrene microspheres with near-identical, spectrally-matched fluorescent protein dye as well as negative control beads in a single dropper bottle.
Specifications
Specifications
Flow Cytometry
Compensation Beads
25 tests
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'