missing translation for 'onlineSavingsMsg'
Learn More

Bromine liquid, 99.8%

CAS: 7726-95-6 | Br | 79.91 g/mol

$95.33 - $1101.55

Chemical Identifiers

CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
MDL Number MFCD00010896
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
PubChem CID 24408
ChEBI CHEBI:29224
SMILES [Br-]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA0090514
View Documents
Thermo Scientific Chemicals
00090514
25 g
Each for $95.33
Only null left
 
AA0090530
View Documents
Thermo Scientific Chemicals
00090530
250 g
Each for $144.23
Only null left
 
AA00905A1
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
000905A1
1 kg
Each for $328.34
Only null left
 
AA00905D9
missing translation for 'viewDocuments'
Thermo Scientific Chemicals
000905D9
5 x 1 kg
Each for $1,101.55
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7726-95-6
79.91
CPELXLSAUQHCOX-UHFFFAOYSA-M
24408
[Br-]
Br
MFCD00010896
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
CHEBI:29224

Specifications

7726-95-6
25 g
Br
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
[Br-]
79.91
CHEBI:29224
99.8%
Bromine
Liquid
99.8%
MFCD00010896
CPELXLSAUQHCOX-UHFFFAOYSA-M
bromide
24408
159.81
3.1g/cm3
Videos
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H330

DOTInformation : Hazard Class: 8; Packaging Group: I

EINECSNumber : 231-778-1

RTECSNumber : EF9100000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only