missing translation for 'onlineSavingsMsg'
Learn More

Bromine liquid, ACS, 99.5%

Maximum level of impurities: Evaporation residue 0.005%, Cl 0.05%, I 0.001%, Sulfur compounds (as S) 0.001%, Heavy Metals (as Pb) 2ppm, Ni 5ppm | CAS: 7726-95-6 | Br | 79.91 g/mol

$78.92 - $2333.74

Chemical Identifiers

CAS 7726-95-6
Molecular Formula Br
Molecular Weight (g/mol) 79.91
MDL Number MFCD00010896
InChI Key CPELXLSAUQHCOX-UHFFFAOYSA-M
Synonym bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
PubChem CID 24408
ChEBI CHEBI:29224
SMILES [Br-]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA3322009
View Documents
Thermo Scientific Chemicals
03322009
10 g
Each for $78.92
Only null left
 
AA3322022
View Documents
Thermo Scientific Chemicals
03322022
100 g
Each for $138.74
Only null left
 
AA3322036
View Documents
Thermo Scientific Chemicals
03322036
500 g
Each for $458.97
Only null left
 
AA3322061
View Documents
Thermo Scientific Chemicals
03322061
5 x 500 g
Each for $2,333.74
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7726-95-6
79.91
CPELXLSAUQHCOX-UHFFFAOYSA-M
24408
[Br-]
Br
MFCD00010896
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
CHEBI:29224

Specifications

7726-95-6
Liquid
99.5%
MFCD00010896
CPELXLSAUQHCOX-UHFFFAOYSA-M
bromide
24408
159.81
ACS
Red-brown fuming liquid
3.12g/cm3 at 20°C
Brown to Red
10 g
Br
bromine, dibromine, brom, bromine solution, brome, bromo, broom, bromine water, bromo italian, bromo spanish
[Br-]
79.91
CHEBI:29224
99.5%
Unpleasant
230 hPa (173mm Hg) at 20°C
Bromine liquid
Videos
Safety and Handling

Safety and Handling

P260-P264b-P271-P280-P284-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H330

DOTInformation : DOT Class: 8; Packing Group: I

EINECSNumber : 231-778-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only