missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bromobenzene, 99%, pure, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 106680025

 View more versions of this product

Catalog No. AC106680025


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Bromobenzene
-31.0°C
1.4900g/mL
51°C
98.5% min. (GC)
C6H5Br
C6H5Br
2.5 L
1.49
benzene, bromo, monobromobenzene, phenyl bromide, bromo-benzene, 1-bromobenzene, phbr, bromo benzene, 4-bromobenzene, unii-co4d5j547l, hsdb 47
QARVLSVVCXYDNA-UHFFFAOYSA-N
bromobenzene
7961
CHEBI:3179
99%
Liquid
108-86-1
Colorless to Yellow
156.0°C
Authentic
Glass bottle
1.5580 to 1.5600
MFCD00000055
05,206
15,1413
Solubility in water: practically insoluble. Other solubilities: 10.4g/100g alcohol (25 c),71.3g/100g ether (25 c),miscible with chloroform and benzene
BrC1=CC=CC=C1
157.01
1.124 mPa.s (20°C)
157.01
Pure

Chemical Identifiers

108-86-1
157.01
QARVLSVVCXYDNA-UHFFFAOYSA-N
7961
bromobenzene
C6H5Br
MFCD00000055
benzene, bromo, monobromobenzene, phenyl bromide, bromo-benzene, 1-bromobenzene, phbr, bromo benzene, 4-bromobenzene, unii-co4d5j547l, hsdb 47
CHEBI:3179
BrC1=CC=CC=C1
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Flammable liquid and vapor.

GHS P Statement
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.

GHS Signal Word: Warning

EINECSNumber : 203-623-8

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only