Learn More
Bromobenzene, 99%, pure
CAS: 108-86-1 | C6H5Br | 157.01 g/mol
$84.75 - $472.67
Chemical Identifiers
| CAS | 108-86-1 |
|---|---|
| Molecular Formula | C6H5Br |
| Molecular Weight (g/mol) | 157.01 |
| MDL Number | MFCD00000055 |
| InChI Key | QARVLSVVCXYDNA-UHFFFAOYSA-N |
| Synonym | benzene, bromo, monobromobenzene, phenyl bromide, bromo-benzene, 1-bromobenzene, phbr, bromo benzene, 4-bromobenzene, unii-co4d5j547l, hsdb 47 |
| PubChem CID | 7961 |
| ChEBI | CHEBI:3179 |
| IUPAC Name | bromobenzene |
| SMILES | BrC1=CC=CC=C1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC106682500
|
Thermo Scientific Chemicals
106682500 |
250 mL | Glass bottle |
Each for $84.75
|
|
||||
|
AC106680010
|
Thermo Scientific Chemicals
106680010 |
1 L | Glass bottle |
Each for $255.48
|
|
||||
|
AC106680025
|
Thermo Scientific Chemicals
106680025 |
2.5 L | Glass bottle |
Each for $472.67
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 108-86-1 | |
| 157.01 | |
| QARVLSVVCXYDNA-UHFFFAOYSA-N | |
| 7961 | |
| bromobenzene |
| C6H5Br | |
| MFCD00000055 | |
| benzene, bromo, monobromobenzene, phenyl bromide, bromo-benzene, 1-bromobenzene, phbr, bromo benzene, 4-bromobenzene, unii-co4d5j547l, hsdb 47 | |
| CHEBI:3179 | |
| BrC1=CC=CC=C1 |
Specifications
| 108-86-1 | |
| Colorless to Yellow | |
| 156.0°C | |
| Authentic | |
| Glass bottle | |
| 1.5580 to 1.5600 | |
| MFCD00000055 | |
| 05,206 | |
| 15,1413 | |
| Solubility in water: practically insoluble. Other solubilities: 10.4g/100g alcohol (25 c),71.3g/100g ether (25 c),miscible with chloroform and benzene | |
| BrC1=CC=CC=C1 | |
| 157.01 | |
| 1.124 mPa.s (20°C) | |
| 157.01 | |
| Pure | |
| Bromobenzene |
| -31.0°C | |
| 1.4900g/mL | |
| 51°C | |
| 98.5% min. (GC) | |
| C6H5Br | |
| C6H5Br | |
| 250 mL | |
| 1.49 | |
| benzene, bromo, monobromobenzene, phenyl bromide, bromo-benzene, 1-bromobenzene, phbr, bromo benzene, 4-bromobenzene, unii-co4d5j547l, hsdb 47 | |
| QARVLSVVCXYDNA-UHFFFAOYSA-N | |
| bromobenzene | |
| 7961 | |
| CHEBI:3179 | |
| 99% | |
| Liquid |
Safety and Handling
GHS H Statement
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Flammable liquid and vapor.
GHS P Statement
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
GHS Signal Word: Warning
EINECSNumber : 203-623-8
RUO – Research Use Only