missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

(Bromomethyl)cyclobutane, 99%, Thermo Scientific™

Chemical Identifiers

CAS 17247-58-4
Molecular Formula C5H9Br
Molecular Weight (g/mol) 149.03
MDL Number MFCD00040960
InChI Key FLHFTXCMKFVKRP-UHFFFAOYSA-N
Synonym bromomethyl cyclobutane, cyclobutylmethyl bromide, cyclobutane, bromomethyl, sfphabikimup@, bromocyclobutylmethane, cyclobutylmethylbromide, bromomethyl-cyclobutane, cydobutyl-methylbromide, bromomethyl cyclobutan
PubChem CID 2734658
IUPAC Name bromomethylcyclobutane
SMILES C1CC(C1)CBr
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC367640050
SDS View Documents
Thermo Scientific Chemicals
367640050
5mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC367640250
SDS View Documents
Thermo Scientific Chemicals
367640250
25mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC367641000
SDS View Documents
Thermo Scientific Chemicals
367641000
100mL Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

17247-58-4
149.03
FLHFTXCMKFVKRP-UHFFFAOYSA-N
2734658
C1CC(C1)CBr
C5H9Br
MFCD00040960
bromomethyl cyclobutane, cyclobutylmethyl bromide, cyclobutane, bromomethyl, sfphabikimup@, bromocyclobutylmethane, cyclobutylmethylbromide, bromomethyl-cyclobutane, cydobutyl-methylbromide, bromomethyl cyclobutan
bromomethylcyclobutane

Specifications

17247-58-4
100.0
145.0°C to 148.0°C
Authentic
Glass bottle
1.4790 to 1.4830
MFCD00040960
05, 22
bromomethyl cyclobutane, cyclobutylmethyl bromide, cyclobutane, bromomethyl, sfphabikimup@, bromocyclobutylmethane, cyclobutylmethylbromide, bromomethyl-cyclobutane, cydobutyl-methylbromide, bromomethyl cyclobutan
FLHFTXCMKFVKRP-UHFFFAOYSA-N
bromomethylcyclobutane
2734658
99%
98.5
1.3390g/mL
27°C
98.5% min. (GC)
C5H9Br
C4H7CH2Br
5mL
1.339
Solubility in water: .. Other solubilities: soluble in alcohols
C1CC(C1)CBr
149.03
149.03
(Bromomethyl)cyclobutane, 99%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only