missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 383850010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution pH 7 | |
7558-79-4,7732-18-5,7487-94-7,7778-77-0,7647-14-5 | |
1 L | |
1 |
Phosphate buffer, traceable to NIST, ready to use | |
1.0000g/mL | |
MFCD03097944 | |
Solubility in water: miscible. |
RUO – Research Use Only