missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use, Thermo Scientific Chemicals
$87.77 - $118.22
Specifications
Chemical Name or Material | Buffer solution pH 7 |
---|---|
Name Note | Phosphate buffer, traceable to NIST, ready to use |
CAS | 7558-79-4,7732-18-5,7487-94-7,7778-77-0,7647-14-5 |
Density | 1.0000g/mL |
MDL Number | MFCD03097944 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AC383855000
|
Thermo Scientific Chemicals
383855000 |
500 mL |
N/A
|
|
|||||
AC383850010
|
Thermo Scientific Chemicals
383850010 |
1 L |
N/A
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Buffer solution pH 7 | |
7558-79-4,7732-18-5,7487-94-7,7778-77-0,7647-14-5 | |
MFCD03097944 | |
Solubility in water: miscible. |
Phosphate buffer, traceable to NIST, ready to use | |
1.0000g/mL | |
1 |
RUO – Research Use Only