missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Buffer solution pH 7, Phosphate buffer, traceable to NIST, ready to use, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 383855000

 View more versions of this product

Catalog No. AC383855000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Buffer solution pH 7
7558-79-4,7732-18-5,7487-94-7,7778-77-0,7647-14-5
500 mL
1
Phosphate buffer, traceable to NIST, ready to use
1.0000g/mL
MFCD03097944
Solubility in water: miscible.
Videos
SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only