Bullard™ Sentinel™ Faceshield Headgear
Lightweight molded ABS plastic headgear offers the ultimate value in face protection.
Supplier:  Bullard™ SENTINEL1
 View more versions of this product
Catalog No. 18-999-3744
Explore available promotions
Description
Description
Standard, lightweight face protection with black ABS crown protector. Easily accommodates primary eye protection and facepiece-style respirators. (For ordering information, contact your Fisher Safety Representative).
Key Features
  • Crown strap conforms to shape of head
  • Hand-washable Sportek™ brow pad absorbs moisture for a drier, more comfortable fit
  • Sure-Lock™ ratchet securely adjusts headband from sizes 6 1/2 to 8, or head circumferences from 20 1/2 to 25 in. (52 to 64cm)
Order Info
Note: Does not include faceshield visor (sold separately).
Compliance
Meets ANSI/ISEA Z87.1 standards.
Specifications
Specifications
6.5 to 8 in.
Sentinel 1 Faceshield Headgear
Black crown protector
Sure-Lock Ratchet
For use with all Bullard visors except 840MX2 (acetate), 840PX2 (polycarbonate) or 840TX2 (Tritan). Visor sold separately.
SDS
Documents
Documents
Accessories