Bulwark™ FR Nomex™ IIIA HRC1 6 oz. Uniform Shirts
Protection: Arc Rating ATPV 5.7 calories/cm2
Supplier:  Bulwark™ FR SND6RBLNL
 View more versions of this product
Catalog No. 19-166-1065
Explore available promotions
Alert:
Not recommended for use around any molten substances, welding operations, or in a critical static control application.
Description
Description
  • Flame-resistant, 6 oz. Nomex IIIA
  • Banded, topstitched collar
  • Topstitched cuffs with adjustable button closures
  • Placket front with button closure
  • Tailored sleeve placket
Specifications
Specifications
Large (Long)
Royal Blue
6 oz.
Nomex IIIA
1