missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bulwark™ FR Nomex™ IIIA HRC1 6 oz. Uniform Shirts
Protection: Arc Rating ATPV 5.7 calories/cm2
$253.55 - $253.55
Specifications
Material | Nomex IIIA |
---|---|
Color | Royal Blue |
Hazard Risk Category | 1 |
Weight (English) | 6 oz. |
Alert:
Not recommended for use around any molten substances, welding operations, or in a critical static control application.
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-166-1062
|
Bulwark™ FR
SND6RBRGXXL |
2X-Large (Regular) |
N/A
|
|
|||||
19-166-1059
|
Bulwark™ FR
SND6RBRGM |
Medium (Regular) |
N/A
|
|
|||||
19-166-1060
|
Bulwark™ FR
SND6RBRGL |
Large (Regular) |
N/A
|
|
|||||
19-166-1061
|
Bulwark™ FR
SND6RBRGXL |
X-Large (Regular) |
N/A
|
|
|||||
19-166-1063
|
Bulwark™ FR
SND6RBRG3XL |
3X-Large (Regular) |
N/A
|
|
|||||
19-166-1064
|
Bulwark™ FR
SND6RBLNM |
Medium (Long) |
N/A
|
|
|||||
19-166-1065
|
Bulwark™ FR
SND6RBLNL |
Large (Long) |
N/A
|
|
|||||
19-166-1067
|
Bulwark™ FR
SND6RBLNXXL |
2X-Large (Long) |
N/A
|
|
|||||
19-166-1066
|
Bulwark™ FR
SND6RBLNXL |
X-Large (Long) |
N/A
|
|
|||||
Description
- Flame-resistant, 6 oz. Nomex IIIA
- Banded, topstitched collar
- Topstitched cuffs with adjustable button closures
- Placket front with button closure
- Tailored sleeve placket
Specifications
Nomex IIIA | |
Royal Blue | |
1 | |
6 oz. |