missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butyl butyrate, 98%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 259590010

 View more versions of this product

Catalog No. AC259590010

Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Butyl butyrate
109-21-7
-92.0°C
0.8675g/mL
49°C
97.5% min. (GC)
C8H16O2
CH3CH2CH2CO2(CH2)3CH3
1 L
0.8675
butyl butyrate, n-butyl butyrate, butanoic acid, butyl ester, n-butyl butanoate, 1-butyl butyrate, butyl butylate, butyric acid butyl ester, n-butyl n-butyrate, butyric acid, butyl ester, butyl n-butyrate
XUPYJHCZDLZNFP-UHFFFAOYSA-N
butyl butanoate
7983
144.2
Liquid
98%
1mg KOH/g max.
Colorless to Yellow
164.0°C to 165.0°C
Authentic
Glass bottle
1.4050 to 1.4070
MFCD00009450
02, 271
15, 1558
Solubility in water: 0.05%. Other solubilities: soluble in alcohol and ether
CCCCOC(=O)CCC
144.21
CHEBI:87429
98%

Chemical Identifiers

109-21-7
144.21
XUPYJHCZDLZNFP-UHFFFAOYSA-N
7983
butyl butanoate
C8H16O2
MFCD00009450
butyl butyrate, n-butyl butyrate, butanoic acid, butyl ester, n-butyl butanoate, 1-butyl butyrate, butyl butylate, butyric acid butyl ester, n-butyl n-butyrate, butyric acid, butyl ester, butyl n-butyrate
CHEBI:87429
CCCCOC(=O)CCC
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-656-8

RTECSNumber : ES8120000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only