missing translation for 'onlineSavingsMsg'
Learn More

Butyl butyrate, 98%

CAS: 109-21-7 | C8H16O2 | 144.21 g/mol

$38.62 - $126.55

Chemical Identifiers

CAS 109-21-7
Molecular Formula C8H16O2
Molecular Weight (g/mol) 144.21
MDL Number MFCD00009450
InChI Key XUPYJHCZDLZNFP-UHFFFAOYSA-N
Synonym butyl butyrate, n-butyl butyrate, butanoic acid, butyl ester, n-butyl butanoate, 1-butyl butyrate, butyl butylate, butyric acid butyl ester, n-butyl n-butyrate, butyric acid, butyl ester, butyl n-butyrate
PubChem CID 7983
ChEBI CHEBI:87429
IUPAC Name butyl butanoate
SMILES CCCCOC(=O)CCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC259591000
SDS View Documents
Thermo Scientific Chemicals
259591000
100 mL Glass bottle
Each for $38.62
Only null left
 
AC259590010
SDS View Documents
Thermo Scientific Chemicals
259590010
1 L Glass bottle
Each for $126.55
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

109-21-7
144.21
XUPYJHCZDLZNFP-UHFFFAOYSA-N
7983
butyl butanoate
C8H16O2
MFCD00009450
butyl butyrate, n-butyl butyrate, butanoic acid, butyl ester, n-butyl butanoate, 1-butyl butyrate, butyl butylate, butyric acid butyl ester, n-butyl n-butyrate, butyric acid, butyl ester, butyl n-butyrate
CHEBI:87429
CCCCOC(=O)CCC

Specifications

109-21-7
-92.0°C
0.8675g/mL
49°C
97.5% min. (GC)
C8H16O2
CH3CH2CH2CO2(CH2)3CH3
100 mL
0.8675
butyl butyrate, n-butyl butyrate, butanoic acid, butyl ester, n-butyl butanoate, 1-butyl butyrate, butyl butylate, butyric acid butyl ester, n-butyl n-butyrate, butyric acid, butyl ester, butyl n-butyrate
XUPYJHCZDLZNFP-UHFFFAOYSA-N
butyl butanoate
7983
144.2
Liquid
1mg KOH/g max.
Colorless to Yellow
164.0°C to 165.0°C
Authentic
Glass bottle
1.4050 to 1.4070
MFCD00009450
02, 271
15, 1558
Solubility in water: 0.05%. Other solubilities: soluble in alcohol and ether
CCCCOC(=O)CCC
144.21
CHEBI:87429
98%
Butyl butyrate, 98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 203-656-8

RTECSNumber : ES8120000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only