Learn More
Butyl butyrate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 259591000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Butyl butyrate | |
109-21-7 | |
-92.0°C | |
0.8675g/mL | |
49°C | |
97.5% min. (GC) | |
C8H16O2 | |
CH3CH2CH2CO2(CH2)3CH3 | |
100 mL | |
0.8675 | |
butyl butyrate, n-butyl butyrate, butanoic acid, butyl ester, n-butyl butanoate, 1-butyl butyrate, butyl butylate, butyric acid butyl ester, n-butyl n-butyrate, butyric acid, butyl ester, butyl n-butyrate | |
XUPYJHCZDLZNFP-UHFFFAOYSA-N | |
butyl butanoate | |
7983 | |
144.2 | |
Liquid |
98% | |
1mg KOH/g max. | |
Colorless to Yellow | |
164.0°C to 165.0°C | |
Authentic | |
Glass bottle | |
1.4050 to 1.4070 | |
MFCD00009450 | |
02, 271 | |
15, 1558 | |
Solubility in water: 0.05%. Other solubilities: soluble in alcohol and ether | |
CCCCOC(=O)CCC | |
144.21 | |
CHEBI:87429 | |
98% |
Chemical Identifiers
109-21-7 | |
144.21 | |
XUPYJHCZDLZNFP-UHFFFAOYSA-N | |
7983 | |
butyl butanoate |
C8H16O2 | |
MFCD00009450 | |
butyl butyrate, n-butyl butyrate, butanoic acid, butyl ester, n-butyl butanoate, 1-butyl butyrate, butyl butylate, butyric acid butyl ester, n-butyl n-butyrate, butyric acid, butyl ester, butyl n-butyrate | |
CHEBI:87429 | |
CCCCOC(=O)CCC |
Safety and Handling
GHS H Statement:
Flammable liquid and vapor.
GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 203-656-8
RTECSNumber : ES8120000
TSCA : TSCA
RUO – Research Use Only