missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butyrylthiocholine Iodide 99.0+%, TCI America™

Supplier:  TCI America B07755G

 View more versions of this product

Catalog No. B07755G

Explore available promotions

Description

Description

Specifications

Specifications

Butyrylthiocholine Iodide
171°C
C9H20INOS
5 g
WEQAAFZDJROSBF-UHFFFAOYSA-M
2-butanoylsulfanylethyl(trimethyl)azanium;iodide
74630
≥99.0% (T)
1866-16-6
White-Yellow
MFCD00011845
butyrylthiocholine iodide, s-butyrylthiocholine iodide, 2-butyrylthio-n,n,n-trimethylethanaminium iodide, s-n-butyrylthiocholine iodide, 2-mercaptoethyl trimethylammonium iodide butyrate, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide, propylcarbonylthioethyl trimethylammonium iodide, 1-2-trimethylamino ethylthio butan-1-one, iodide, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide 1:1
CCCC(=O)SCC[N+](C)(C)C.[I-]
317.229
317.23
Crystalline Powder

Chemical Identifiers

1866-16-6
317.229
WEQAAFZDJROSBF-UHFFFAOYSA-M
74630
CCCC(=O)SCC[N+](C)(C)C.[I-]
C9H20INOS
MFCD00011845
butyrylthiocholine iodide, s-butyrylthiocholine iodide, 2-butyrylthio-n,n,n-trimethylethanaminium iodide, s-n-butyrylthiocholine iodide, 2-mercaptoethyl trimethylammonium iodide butyrate, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide, propylcarbonylthioethyl trimethylammonium iodide, 1-2-trimethylamino ethylthio butan-1-one, iodide, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide 1:1
2-butanoylsulfanylethyl(trimethyl)azanium;iodide
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions