missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Butyrylthiocholine Iodide 99.0+%, TCI America™

$132.32 - $132.32

Chemical Identifiers

CAS 1866-16-6
Molecular Formula C9H20INOS
Molecular Weight (g/mol) 317.229
MDL Number MFCD00011845
InChI Key WEQAAFZDJROSBF-UHFFFAOYSA-M
Synonym butyrylthiocholine iodide, s-butyrylthiocholine iodide, 2-butyrylthio-n,n,n-trimethylethanaminium iodide, s-n-butyrylthiocholine iodide, 2-mercaptoethyl trimethylammonium iodide butyrate, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide, propylcarbonylthioethyl trimethylammonium iodide, 1-2-trimethylamino ethylthio butan-1-one, iodide, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide 1:1
PubChem CID 74630
IUPAC Name 2-butanoylsulfanylethyl(trimethyl)azanium;iodide
SMILES CCCC(=O)SCC[N+](C)(C)C.[I-]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B07755G
View Documents
TCI America
B07755G
5 g
Each for $132.32
 
Description

Description

Specifications

Chemical Identifiers

1866-16-6
317.229
WEQAAFZDJROSBF-UHFFFAOYSA-M
74630
CCCC(=O)SCC[N+](C)(C)C.[I-]
C9H20INOS
MFCD00011845
butyrylthiocholine iodide, s-butyrylthiocholine iodide, 2-butyrylthio-n,n,n-trimethylethanaminium iodide, s-n-butyrylthiocholine iodide, 2-mercaptoethyl trimethylammonium iodide butyrate, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide, propylcarbonylthioethyl trimethylammonium iodide, 1-2-trimethylamino ethylthio butan-1-one, iodide, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide 1:1
2-butanoylsulfanylethyl(trimethyl)azanium;iodide

Specifications

1866-16-6
White-Yellow
MFCD00011845
butyrylthiocholine iodide, s-butyrylthiocholine iodide, 2-butyrylthio-n,n,n-trimethylethanaminium iodide, s-n-butyrylthiocholine iodide, 2-mercaptoethyl trimethylammonium iodide butyrate, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide, propylcarbonylthioethyl trimethylammonium iodide, 1-2-trimethylamino ethylthio butan-1-one, iodide, ethanaminium, n,n,n-trimethyl-2-1-oxobutyl thio-, iodide 1:1
CCCC(=O)SCC[N+](C)(C)C.[I-]
317.229
317.23
Crystalline Powder
171°C
C9H20INOS
5 g
WEQAAFZDJROSBF-UHFFFAOYSA-M
2-butanoylsulfanylethyl(trimethyl)azanium;iodide
74630
≥99.0% (T)
Butyrylthiocholine Iodide
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'