Innovatek C-Reactive Protein (CPR) Latex Test Kits
Innovatek C-Reactive Protein (CPR) Latex Test Kits offer results in less than 3 minutes
Supplier: Innovatek 419110102
Description
- Fast 2 to 3 minute rotation
- Complete kit includes all reagents and controls
- Disposable test cards
- 6 to 12 months of shelf life
- Sensitive and specific
Specifications
C-Reactive Protein (CRP) | |
Single-use | |
50 Tests/Kit | |
≈3 min. |