Innovatek C-Reactive Protein (CPR) Latex Test Kits
Innovatek C-Reactive Protein (CPR) Latex Test Kits offer results in less than 3 minutes
$125.61 - $164.97
Specifications
Detectable Analytes | C-Reactive Protein (CRP) |
---|---|
Disposable | Single-use |
Test Time | ≈3 min. |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
29-801-065
|
Innovatek
419110102 |
50 Tests/Kit |
Pack of 50 Each for $125.61
|
|
|||||
29-801-066
|
Innovatek
419110103 |
100 Tests/Kit |
Pack of 100 Each for $164.97
|
|
|||||
Description
- Fast 2 to 3 minute rotation
- Complete kit includes all reagents and controls
- Disposable test cards
- 6 to 12 months of shelf life
- Sensitive and specific
Specifications
C-Reactive Protein (CRP) | |
Single-use | |
≈3 min. |