Cadence Science™ Laboratory Pipetting Needles
316 Stainless Steel Hub with 316 Stainless Steel Tubing
Supplier: Cadence Science™ POP7940
Description
316 Stainless Steel Hub with 316 Stainless Steel Tubing
- Needles 14-825-16A through -16N and 14-818-106A through -106C all have standard nickel plated hubs and 304 stainless steel cannulae
- Needles 01-290-29 through -35 have 316 stainless steel Micro-Mate™ Threaded Female Luer Hubs and 316 stainless steel cannulae
- All have 90° blunt ends
Specifications
Nickel-plated (Hub), Stainless Steel | |
4 in. | |
12 Needles | |
Manual and Automatic Volumetric Dispensers, Diluters, Pipetters |
15 G | |
Pipetting Needle | |
90° Blunt | |
Luer |