Cadence Science™ Laboratory Pipetting Needles
316 Stainless Steel Hub with 316 Stainless Steel Tubing
$778.00 - $2293.61
Catalog Number | Mfr. No. | Needle Gauge | Length (English) Needle | Material | Connection Type | Price | Quantity | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Needle Gauge | Length (English) Needle | Material | Connection Type | Price | Quantity | ||
14818106A
|
Cadence Science™
7932 |
20 G | 1.5 in. | Nickel-plated (Hub), Stainless Steel | Luer |
Dozen of 12 Each for $778.00
|
|
||
0129029
|
Cadence Science™
9870 |
14 G | 4 in. | Stainless Steel, Stainless Steel (Hub) | Female Luer |
Dozen of 12 Each for $2,293.61
|
|
||
01-290-30
|
Cadence Science™
POP9871 |
14 G | 6 in. | Stainless Steel, Stainless Steel (Hub) | Female Luer |
Dozen of 12 Each for $2,226.15
|
|
||
01-290-32
|
Cadence Science™
9874 |
18 G | 2 in. | Stainless Steel, Stainless Steel (Hub) | Female Luer |
Dozen of 12 Each for $2,293.61
|
|
||
14-818-106C
|
Cadence Science™
POP7940 |
15 G | 4 in. | Nickel-plated (Hub), Stainless Steel | Luer |
Dozen of 12 Each for $778.00
|
|
||
14-825-16A
|
Cadence Science™
POP7930 |
24 G | 1 in. | Nickel-plated (Hub), Stainless Steel | Luer |
Dozen of 12 Each for $778.00
|
|
||
14-825-16B
|
Cadence Science™
POP7931 |
22 G | 1.5 in. | Nickel-plated (Hub), Stainless Steel | Luer |
Dozen of 12 Each for $778.00
|
|
||
14-825-16F
|
Cadence Science™
POP7935 |
18 G | 2 in. | Nickel-plated (Hub), Stainless Steel | Luer |
Dozen of 12 Each for $778.00
|
|
||
14-825-16M
|
Cadence Science™
POP7941 |
14 G | 4 in. | Nickel-plated (Hub), Stainless Steel | Luer |
Dozen of 12 Each for $844.91
|
|
||
14-825-16N
|
Cadence Science™
POP7942 |
14 G | 6 in. | Nickel-plated (Hub), Stainless Steel | Luer |
Dozen of 12 Each for $844.91
|
|
||
14-825-16G
|
Cadence Science™
POP7936 |
18 G | 4 in. | Nickel-plated (Hub), Stainless Steel | Luer |
Dozen of 12 Each for $778.00
|
|
||
14-825-16E
|
Cadence Science™
POP7934 |
20 G | 6 in. | Nickel-plated (Hub), Stainless Steel | Luer |
Dozen of 12 Each for $844.91
|
|
||
14-825-16H
|
Cadence Science™
POP7937 |
18 G | 6 in. | Nickel-plated (Hub), Stainless Steel | Luer |
Dozen of 12 Each for $844.91
|
|
||
14-825-16J
|
Cadence Science™
POP7938 |
16 G | 2 in. | Nickel-plated (Hub), Stainless Steel | Luer |
Dozen of 12 Each for $778.00
|
|
||
14-825-16K
|
Cadence Science™
POP7939 |
16 G | 4 in. | Nickel-plated (Hub), Stainless Steel | Luer |
Dozen of 12 Each for $844.91
|
|
||
Description
316 Stainless Steel Hub with 316 Stainless Steel Tubing
- Needles 14-825-16A through -16N and 14-818-106A through -106C all have standard nickel plated hubs and 304 stainless steel cannulae
- Needles 01-290-29 through -35 have 316 stainless steel Micro-Mate™ Threaded Female Luer Hubs and 316 stainless steel cannulae
- All have 90° blunt ends
Specifications
Pipetting Needle | |
90° Blunt |