Cadence Science™ Laboratory Pipetting Needles
316 Stainless Steel Hub with 316 Stainless Steel Tubing
Supplier:  Cadence Science™ POP7930
 View more versions of this product
Catalog No. 14-825-16A
Explore available promotions
Description
Description
316 Stainless Steel Hub with 316 Stainless Steel Tubing
  • Needles 14-825-16A through -16N and 14-818-106A through -106C all have standard nickel plated hubs and 304 stainless steel cannulae
  • Needles 01-290-29 through -35 have 316 stainless steel Micro-Mate™ Threaded Female Luer Hubs and 316 stainless steel cannulae
  • All have 90° blunt ends
Specifications
Specifications
Nickel-plated (Hub), Stainless Steel
For both manual and automatic volumetric dispensers, diluters, pipetters and for laboratory procedures
Pipetting Needle
90° Blunt
24 G
1 in.
12 Needles
Luer