missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cadence Science™ Reusable Small Animal Feeding Needles: Curved

Precision-assembled stainless-steel feeding needles ideal for feeding and dosing

Supplier:  Cadence Science™ POP7914

 View more versions of this product

Catalog No. 01-290-10C


Explore available promotions

Description

Description

Precision-assembled stainless-steel feeding needles ideal for feeding and dosing

  • For the intubation of laboratory animals
  • Straight, curved tubing styles
  • Many standard sizes available, from small to large
  • Curved
  • Autoclavable

Small Animal Needles

  • For animals 0.7 oz. to 1 lb., 5 oz. (20 to 600g)

Specifications

Specifications

Curved Animal Feeding Needle
Stainless Steel
Small animal, ideal for feeding and dosing
3 in.
0.088 in.
Autoclavable
Reusable
18G
0.76 cm
0.023 cm
Videos
SDS
Documents

Documents

Promotions

Promotions