missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium acetate monohydrate, ACS reagent, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 446355000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Calcium acetate monohydrate | |
5743-26-0 | |
C4H6CaO4·H2O | |
15,1649 | |
Solubility in water: soluble | |
CC(=O)[O-].CC(=O)[O-].O.[Ca+2] | |
176.19 | |
CHEBI:59199 | |
ACS Reagent |
160.0°C | |
500 g | |
(CH3CO2)2Ca·H2O | |
calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu | |
XQKKWWCELHKGKB-UHFFFAOYSA-L | |
calcium;diacetate;hydrate | |
82163 | |
176.19 | |
Plastic bottle |
Chemical Identifiers
5743-26-0 | |
176.19 | |
calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu | |
CHEBI:59199 | |
CC(=O)[O-].CC(=O)[O-].O.[Ca+2] |
C4H6CaO4·H2O | |
XQKKWWCELHKGKB-UHFFFAOYSA-L | |
82163 | |
calcium;diacetate;hydrate |
RUO – Research Use Only