missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium acetate monohydrate, ACS reagent, Thermo Scientific Chemicals
$320.34 - $320.34
Chemical Identifiers
CAS | 5743-26-0 |
---|---|
Molecular Formula | C4H6CaO4·H2O |
Molecular Weight (g/mol) | 176.19 |
InChI Key | XQKKWWCELHKGKB-UHFFFAOYSA-L |
Synonym | calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu |
PubChem CID | 82163 |
ChEBI | CHEBI:59199 |
IUPAC Name | calcium;diacetate;hydrate |
SMILES | CC(=O)[O-].CC(=O)[O-].O.[Ca+2] |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC446355000
|
Thermo Scientific Chemicals
446355000 |
500 g | Plastic bottle |
Each for $320.34
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5743-26-0 | |
176.19 | |
calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu | |
CHEBI:59199 | |
CC(=O)[O-].CC(=O)[O-].O.[Ca+2] |
C4H6CaO4·H2O | |
XQKKWWCELHKGKB-UHFFFAOYSA-L | |
82163 | |
calcium;diacetate;hydrate |
Specifications
160.0°C | |
500 g | |
(CH3CO2)2Ca·H2O | |
calcium acetate monohydrate, calcium diacetate monohydrate, unii-7za48gim5h, calcium diacetate hydrate, acetic acid, calcium salt, monohydrate, calcium acetate, monohydrate, 7za48gim5h, calcium acetat, calcium acetate jan, acmc-1c5pu | |
XQKKWWCELHKGKB-UHFFFAOYSA-L | |
calcium;diacetate;hydrate | |
82163 | |
176.19 | |
Plastic bottle |
5743-26-0 | |
C4H6CaO4·H2O | |
15,1649 | |
Solubility in water: soluble | |
CC(=O)[O-].CC(=O)[O-].O.[Ca+2] | |
176.19 | |
CHEBI:59199 | |
ACS Reagent | |
Calcium acetate monohydrate |
RUO – Research Use Only