Learn More
Calcium chloride, 93%, extra pure, powder, anhydrous, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 349615000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
AdsorbentSpecifications
>1600.0°C | |
782.0°C | |
10043-52-4 | |
Powder | |
95% min. (Complexometry) | |
CaCl2 | |
01,104; 04,368 | |
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal | |
UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
calcium;dichloride | |
5284359 | |
110.99 | |
Extra Pure |
Calcium chloride | |
96% | |
White | |
500 g | |
CaCl2 | |
MFCD00010903 | |
15,1661 | |
Solubility in water: 740g/L (20°C). Other solubilities: freely soluble in alcohol | |
[Cl-].[Cl-].[Ca++] | |
110.98 | |
CHEBI:3312 | |
96% | |
Plastic bottle |
Chemical Identifiers
10043-52-4 | |
110.98 | |
UXVMQQNJUSDDNG-UHFFFAOYSA-L | |
5284359 | |
[Cl-].[Cl-].[Ca++] |
CaCl2 | |
MFCD00010903 | |
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal | |
CHEBI:3312 |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 233-140-8
RTECSNumber : EV9800000
TSCA : TSCA
RUO – Research Use Only