missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium chloride, 93%, extra pure, powder, anhydrous, Thermo Scientific Chemicals

$37.22 - $1263.79

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
MDL Number MFCD00010903
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
PubChem CID 5284359
ChEBI CHEBI:3312
SMILES [Cl-].[Cl-].[Ca++]
View More Specs

Products 4
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC349610250
SDS View Documents
Thermo Scientific Chemicals
349610250
25 g Plastic bottle
Each for $37.22
 
AC349615000
SDS View Documents
Thermo Scientific Chemicals
349615000
500 g Plastic bottle
Each for $127.19
 
AC349610025
SDS View Documents
Thermo Scientific Chemicals
349610025
2.5 kg Plastic bottle
Each for $433.89
 
AC349610100
SDS View Documents
Thermo Scientific Chemicals
349610100
10 kg Plastic drum
Each for $1,263.79
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Adsorbent
Specifications

Chemical Identifiers

10043-52-4
110.98
UXVMQQNJUSDDNG-UHFFFAOYSA-L
5284359
[Cl-].[Cl-].[Ca++]
CaCl2
MFCD00010903
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
CHEBI:3312

Specifications

>1600.0°C
10043-52-4
Powder
95% min. (Complexometry)
CaCl2
01,104; 04,368
calcium chloride, calcium dichloride, calcium chloride anhydrous, calcium ii chloride, calciumchloride, caloride, liquical, calcium chloride pellets, isocal, jarcal
UXVMQQNJUSDDNG-UHFFFAOYSA-L
calcium dichloride
5284359
110.99
Extra Pure
Calcium chloride, 96%
782.0°C
White
25 g
CaCl2
MFCD00010903
15,1661
Solubility in water: 740g/L (20°C). Other solubilities: freely soluble in alcohol
[Cl-].[Cl-].[Ca++]
110.98
CHEBI:3312
96%
Plastic bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 233-140-8

RTECSNumber : EV9800000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only