missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium Chloride Dihydrate (Certified ACS), Fisher Chemical™

Supplier:  Thermo Fisher Scientific FLC793

 View more versions of this product

Catalog No. C79-3



Explore available promotions

Description

Description

Specifications

Specifications

Calcium Chloride Dihydrate
Beige-White
4.5 to 6.5
3 kg
Ca Cl2 · 2 H2O
147.01
Certified ACS
Poly Bottle
175°C
10035-04-8
Solid
99.0 to 105.0 %
MFCD00149613
99.0 to 105.0%
Pass test
0.830g/cm³

Chemical Identifiers

10035-04-8
Ca Cl2 · 2 H2O
MFCD00149613
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Irritating to eyes. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:1

SDS Documents

Documents

Promotions

Promotions