missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium Chloride Dihydrate (Certified ACS), Fisher Chemical™

$195.00 - $894.42

Chemical Identifiers

CAS 10035-04-8
Molecular Formula Ca Cl2 · 2 H2O
MDL Number MFCD00149613
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
C79-500 Edge
SDS View Documents Promotion Details
Fisher Chemical
FLC79500
500 g Poly Bottle
Each for $195.00
Case of 6 Each for $1,170.00
Edge
 
C79-3
SDS View Documents Promotion Details
Thermo Fisher Scientific
FLC793
3 kg Poly Bottle
Each for $894.42
Case of 4 Each for $3,577.68
 
Description

Description

Specifications

Chemical Identifiers

10035-04-8
Ca Cl2 · 2 H2O
MFCD00149613

Specifications

175°C
10035-04-8
Solid
99.0 to 105.0 %
MFCD00149613
99.0 to 105.0%
Pass test
0.830g/cm³
Beige-White
4.5 to 6.5
500 g
Ca Cl2 · 2 H2O
147.01
Certified ACS
Poly Bottle
Calcium Chloride Dihydrate
Videos
Safety and Handling

Safety and Handling

WARNING!

Emergency Overview
Irritating to eyes. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:2
Flammability:0
Instability:1

SDS
missing translation for 'documents'

missing translation for 'documents'