missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Calcium Chloride Dihydrate (Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical FLC79500
Specifications
Calcium Chloride Dihydrate | |
Beige-White | |
4.5 to 6.5 | |
500 g | |
Ca Cl2 · 2 H2O | |
147.01 | |
Certified ACS | |
Poly Bottle |
175°C | |
10035-04-8 | |
Solid | |
99.0 to 105.0 % | |
MFCD00149613 | |
99.0 to 105.0% | |
Pass test | |
0.830g/cm³ |
Chemical Identifiers
10035-04-8 | |
Ca Cl2 · 2 H2O | |
MFCD00149613 |
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:2
Flammability:0
Instability:1