missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium chloride hexahydrate, 98+%, for analysis, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 389262500

 View more versions of this product

Catalog No. AC389262500


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Calcium chloride hexahydrate
For analysis
Colorless
Chunks
98% min. (Complexometry)
CaCl2·6H2O
15, 1661
Solubility in water: 5360g/L (20°C)
O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2]
219.08
219.08
Analytical
30.0°C
7774-34-7
4.5 to 8.5 (5% soln.)
250 g
CaCl2·6H2O
MFCD00149616
calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g
QHFQAJHNDKBRBO-UHFFFAOYSA-L
calcium;dichloride;hexahydrate
6093252
≥98%
Plastic bottle

Chemical Identifiers

7774-34-7
219.08
QHFQAJHNDKBRBO-UHFFFAOYSA-L
6093252
O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2]
CaCl2·6H2O
MFCD00149616
calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g
calcium;dichloride;hexahydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only