missing translation for 'onlineSavingsMsg'
Learn More

Calcium chloride hexahydrate, 98+%, for analysis

CAS: 7774-34-7 | CaCl2·6H2O | 219.08 g/mol

$55.81 - $208.05

Chemical Identifiers

CAS 7774-34-7
Molecular Formula CaCl2·6H2O
Molecular Weight (g/mol) 219.08
MDL Number MFCD00149616
InChI Key QHFQAJHNDKBRBO-UHFFFAOYSA-L
Synonym calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g
PubChem CID 6093252
IUPAC Name calcium;dichloride;hexahydrate
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2]
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC389262500
SDS View Documents
Thermo Scientific Chemicals
389262500
250 g Plastic bottle
Each for $55.81
Only null left
 
AC389260010
SDS View Documents
Thermo Scientific Chemicals
389260010
1 kg Plastic bottle
Each for $114.01
Only null left
 
AC389260025
SDS View Documents
Thermo Scientific Chemicals
389260025
2.5 kg Plastic bottle
Each for $208.05
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7774-34-7
219.08
QHFQAJHNDKBRBO-UHFFFAOYSA-L
6093252
O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2]
CaCl2·6H2O
MFCD00149616
calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g
calcium;dichloride;hexahydrate

Specifications

30.0°C
Colorless
Chunks
98% min. (Complexometry)
CaCl2·6H2O
15, 1661
Solubility in water: 5360g/L (20°C)
O.O.O.O.O.O.[Cl-].[Cl-].[Ca+2]
219.08
219.08
Analytical
Calcium chloride hexahydrate, For analysis
7774-34-7
4.5 to 8.5 (5% soln.)
250 g
CaCl2·6H2O
MFCD00149616
calcium chloride hexahydrate, antarcticite, acmc-20alis, calcium hexahydrate dichloride, calcium chloride-water 1/6, calcium chloride hydrate, puratronic, calcium chloride, hydrated, calcium chloride hydrate trace metals basis 1g
QHFQAJHNDKBRBO-UHFFFAOYSA-L
calcium;dichloride;hexahydrate
6093252
≥98%
Plastic bottle
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only