missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Calcium citrate tetrahydrate, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1482836

 View more versions of this product

Catalog No. AAA1482836


Explore available promotions

Description

Description

Calcium citrate tetrahydrate may be used as raw material in the preparation of nanoparticle with the nominal composition Ca0.8 Ba0.2 Ti03 :Pr3+, via sol-gel process. It may be employed as carbon source in a simple and effective carbonization method to prepare porous carbon without any activation process. It is suitable for use in the preparation of bovine serum albumin (BSA) nanoparticles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Calcium citrate tetrahydrate may be used as raw material in the preparation of nanoparticle with the nominal composition Ca0.8 Ba0.2 Ti03 :Pr3+, via sol-gel process. It may be employed as carbon source in a simple and effective carbonization method to prepare porous carbon without any activation process. It is suitable for use in the preparation of bovine serum albumin (BSA) nanoparticles.

Solubility
Soluble in water and hydrochloric acid.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

Calcium citrate tetrahydrate
C12H18Ca3O18
500 g
Soluble in water and hydrochloric acid.
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O.[Ca+2].[Ca+2].[Ca+2]
570.492
572.60 (500.54 Anhydrous)
5785-44-4
MFCD00150786
calcium citrate tetrahydrate, unii-mlm29u2x85, calcium citrate usp, tricalcium dicitrate tetrahydrate, citric acid calcium salt, citracal tn, calcium citrate 3:2 , tetrahydrate, acmc-20aliq, ksc493e4f
LNIZKKFWMDARJV-UHFFFAOYSA-H
tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate
5282392
96%

Chemical Identifiers

5785-44-4
570.492
LNIZKKFWMDARJV-UHFFFAOYSA-H
5282392
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O.[Ca+2].[Ca+2].[Ca+2]
C12H18Ca3O18
MFCD00150786
calcium citrate tetrahydrate, unii-mlm29u2x85, calcium citrate usp, tricalcium dicitrate tetrahydrate, citric acid calcium salt, citracal tn, calcium citrate 3:2 , tetrahydrate, acmc-20aliq, ksc493e4f
tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only