Learn More
Calcium citrate tetrahydrate, 96%, Thermo Scientific Chemicals
$87.48 - $181.16
Chemical Identifiers
CAS | 5785-44-4 |
---|---|
Molecular Formula | C12H18Ca3O18 |
Molecular Weight (g/mol) | 570.492 |
MDL Number | MFCD00150786 |
InChI Key | LNIZKKFWMDARJV-UHFFFAOYSA-H |
Synonym | calcium citrate tetrahydrate, unii-mlm29u2x85, calcium citrate usp, tricalcium dicitrate tetrahydrate, citric acid calcium salt, citracal tn, calcium citrate 3:2 , tetrahydrate, acmc-20aliq, ksc493e4f |
PubChem CID | 5282392 |
IUPAC Name | tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate |
SMILES | C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O.[Ca+2].[Ca+2].[Ca+2] |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
AAA1482822
|
Thermo Scientific Chemicals
A1482822 |
100 g |
Each for $87.48
|
|
||||||
AAA1482836
|
Thermo Scientific Chemicals
A1482836 |
500 g |
Each for $181.16
|
|
||||||
Description
Calcium citrate tetrahydrate may be used as raw material in the preparation of nanoparticle with the nominal composition Ca0.8 Ba0.2 Ti03 :Pr3+, via sol-gel process. It may be employed as carbon source in a simple and effective carbonization method to prepare porous carbon without any activation process. It is suitable for use in the preparation of bovine serum albumin (BSA) nanoparticles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsCalcium citrate tetrahydrate may be used as raw material in the preparation of nanoparticle with the nominal composition Ca0.8 Ba0.2 Ti03 :Pr3+, via sol-gel process. It may be employed as carbon source in a simple and effective carbonization method to prepare porous carbon without any activation process. It is suitable for use in the preparation of bovine serum albumin (BSA) nanoparticles.
Solubility
Soluble in water and hydrochloric acid.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Chemical Identifiers
5785-44-4 | |
570.492 | |
LNIZKKFWMDARJV-UHFFFAOYSA-H | |
5282392 | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O.[Ca+2].[Ca+2].[Ca+2] |
C12H18Ca3O18 | |
MFCD00150786 | |
calcium citrate tetrahydrate, unii-mlm29u2x85, calcium citrate usp, tricalcium dicitrate tetrahydrate, citric acid calcium salt, citracal tn, calcium citrate 3:2 , tetrahydrate, acmc-20aliq, ksc493e4f | |
tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate |
Specifications
5785-44-4 | |
MFCD00150786 | |
calcium citrate tetrahydrate, unii-mlm29u2x85, calcium citrate usp, tricalcium dicitrate tetrahydrate, citric acid calcium salt, citracal tn, calcium citrate 3:2 , tetrahydrate, acmc-20aliq, ksc493e4f | |
LNIZKKFWMDARJV-UHFFFAOYSA-H | |
tricalcium;2-hydroxypropane-1,2,3-tricarboxylate;tetrahydrate | |
5282392 | |
96% |
C12H18Ca3O18 | |
100 g | |
Soluble in water and hydrochloric acid. | |
C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.C(C(=O)[O-])C(CC(=O)[O-])(C(=O)[O-])O.O.O.O.O.[Ca+2].[Ca+2].[Ca+2] | |
570.492 | |
572.60 (500.54 Anhydrous) | |
Calcium citrate tetrahydrate |
Safety and Handling
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only